Assay Details
| Gene Symbol: Prss21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020487.4 | NP_065233.2 | 5 - 6 | 761 | 101 | |
| GenBank mRNA | AB033822.1 | - | 5 - 6 | 747 | 101 | |
| AB059415.1 | - | 5 - 6 | 759 | 101 | ||
| AF176209.2 | - | 5 - 6 | 761 | 101 | ||
| AK006271.1 | - | 5 - 6 | 733 | 101 | ||
| AY005145.1 | - | 5 - 6 | 738 | 101 | ||
| BC049588.1 | - | 5 - 6 | 775 | 101 | ||
| BC116755.1 | - | 5 - 6 | 759 | 40126074 | 101 | |
| BC116757.1 | - | 5 - 6 | 759 | 40126076 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473308 | BC049588.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473308 | BC116755.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473308 | BC116757.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473308 | AB059415.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473308 | AF176209.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473308 | BC049588.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473308 | BC116755.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473308 | BC116757.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473308 | AB059415.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473308 | AF176209.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918999 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780971 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120812 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473308 | BC049588.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473308 | BC116755.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473308 | BC116757.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473308 | AB059415.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473308 | AF176209.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473308 | BC049588.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473308 | BC116755.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473308 | BC116757.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473308 | AB059415.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473308 | AF176209.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473308 | BC049588.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473308 | BC116755.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473308 | BC116757.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473308 | AB059415.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473308 | AF176209.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473308 | BC049588.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473308 | BC116755.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473308 | BC116757.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473308 | AB059415.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473308 | AF176209.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094105 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120812 | - | - |
| Whole Mouse Genome G4122A | A_51_P473308 | BC049588.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473308 | BC116755.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473308 | BC116757.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473308 | AB059415.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473308 | AF176209.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473308 | BC049588.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473308 | BC116755.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473308 | BC116757.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473308 | AB059415.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473308 | AF176209.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137620 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127114 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120812 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13251389 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13251380 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13251381 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13251372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13212158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13166188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13251361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13388172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13345542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13251363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13251378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13206040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13157042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13166187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13251389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13527973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13108192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094116 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442366 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442368 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442367 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442364 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442369 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442365 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419623_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442363 | - | - |
| Mouse Genome 430 2.0 Array | 1419623_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419623_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57256_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57256_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700002392.mm.1 | NM_020487.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002392.mm.1 | NM_020487.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002392.mm.1 | TC1700000326.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002393.mm.1 | NM_020487.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002393.mm.1 | NM_020487.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002393.mm.1 | TC1700000326.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419623_PM_at | - | - |
| Mouse Expression Set 430 | 1419623_at | - | - |
| Murine Genome U74v2 Set | 134119_PM_at | - | - |
| Murine Genome U74v2 Set | 134119_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419623_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419623_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419623_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419623_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6130619 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6130619 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006130619 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2661685 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2661685 | - | - |
| mouse-6 v1.1 (Illumina) | scl51027.6.1_221-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6130619 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl51027.6.1_221-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6130619 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2661685 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2661685 | - | - |

Prss21
Genomic Map