Assay Details
| Gene Symbol: Ttc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033149.2 | NP_001028321.1 | 1 - 2 | 436 | 87 | |
| GenBank mRNA | AK133456.1 | - | 1 - 2 | 437 | 87 | |
| AK161499.1 | - | 1 - 2 | 236 | 87 | ||
| AK172918.1 | - | 556 | 87 | |||
| BC028891.1 | - | 1 - 2 | 520 | 87 | ||
| BC139242.1 | - | 1 - 2 | 434 | 8862264 | 87 | |
| BC139244.1 | - | 1 - 2 | 434 | 8862266 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc9
Genomic Map