Assay Details
| Gene Symbol: Htr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008311.2 | NP_032337.2 | 1 - 2 | 95 | 86 | |
| GenBank mRNA | AK033713.1 | - | 1 - 2 | 95 | 86 | |
| BC023690.1 | - | 1 - 2 | 44 | 5344328 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033713.1, BC023690.1, NM_008311.2 transcripts.
Set Membership: |
|

Htr2b
Genomic Map