Assay Details
| Gene Symbol: 1700012P22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027056.1 | NP_081332.1 | 3 - 4 | 581 | 71 | |
| GenBank mRNA | AK005933.1 | - | 3 - 4 | 582 | 71 | |
| BC099569.1 | - | 3 - 4 | 581 | 30918007 | 71 | |
| BC116372.1 | - | 3 - 4 | 507 | 40100233 | 71 | |
| BC126971.1 | - | 3 - 4 | 507 | 40100231 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139349 | BC116372.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139349 | BC126971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139349 | NM_027056.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139349 | AK005933.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139349 | BC099569.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139349 | BC116372.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139349 | BC126971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139349 | NM_027056.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139349 | AK005933.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139349 | BC099569.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 761778 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303044 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724027 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125450 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 582118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229283 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137388 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 202085 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 746003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 834711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 938818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 248925 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46124 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41797 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136353 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139349 | BC116372.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139349 | BC126971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139349 | NM_027056.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139349 | AK005933.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139349 | BC099569.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139349 | BC116372.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139349 | BC126971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139349 | NM_027056.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139349 | AK005933.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139349 | BC099569.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01011316 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136353 | - | - |
| Whole Mouse Genome G4122A | A_51_P139349 | BC116372.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139349 | BC126971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139349 | NM_027056.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139349 | AK005933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139349 | BC099569.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139349 | BC116372.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139349 | BC126971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139349 | NM_027056.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139349 | AK005933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139349 | BC099569.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1005096 | BC116372.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1005096 | BC126971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1005096 | NM_027056.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1005096 | AK005933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1005096 | BC099569.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03319546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03319545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03377599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03769988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03266479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03801165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03769985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03705360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23378491 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10518211 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518212 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518214 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518213 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518215 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518210 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443845_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453333_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10518209 | - | - |
| Mouse Genome 430 2.0 Array | 1443845_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1453333_at | - | - |
| Mouse Genome 430 2.0 Array | 1443845_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1453333_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69364_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69364_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400032658.mm.1 | NM_027056.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400032658.mm.1 | NM_027056.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400032658.mm.1 | TC0400003943.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400032656.mm.1 | NM_027056.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400032656.mm.1 | NM_027056.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400032656.mm.1 | TC0400003943.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453333_at | - | - |
| Mouse Expression Set 430 | 1443845_PM_x_at | - | - |
| Mouse Expression Set 430 | 1443845_x_at | - | - |
| Mouse Expression Set 430 | 1453333_PM_at | - | - |
| Murine Genome U74v2 Set | 169310_PM_r_at | - | - |
| Murine Genome U74v2 Set | 132022_PM_f_at | - | - |
| Murine Genome U74v2 Set | 169310_r_at | - | - |
| Murine Genome U74v2 Set | 132022_f_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453333_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443845_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453333_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443845_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 582303 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229341 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229341 | - | - |

1700012P22Rik
Genomic Map