Assay Details
| Gene Symbol: Pigq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291025.1 | NP_001277954.1 | 9 - 10 | 1786 | 66 | |
| NM_011822.4 | NP_035952.2 | 8 - 9 | 1606 | 66 | ||
| GenBank mRNA | AK155173.1 | - | 7 - 8 | 1787 | 66 | |
| AK163680.1 | - | 8 - 9 | 1582 | 66 | ||
| AK169284.1 | - | 8 - 9 | 1589 | 66 | ||
| AK170431.1 | - | 9 - 10 | 1733 | 66 | ||
| BC003917.1 | - | 6 - 7 | 696 | 66 | ||
| BC014287.1 | - | 8 - 9 | 1579 | 4193343 | 66 | |
More Information
Set Membership: |
|

Pigq
Genomic Map