Assay Details
| Gene Symbol: Nos2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010927.3 | NP_035057.1 | 5 - 6 | 627 | 82 | |
| GenBank mRNA | AF065919.1 | - | 5 - 6 | 624 | 82 | |
| AF065920.1 | - | 5 - 6 | 624 | 82 | ||
| AF065921.2 | - | 5 - 6 | 624 | 82 | ||
| AF065922.2 | - | 4 - 5 | 497 | 82 | ||
| AF065923.2 | - | 4 - 5 | 497 | 82 | ||
| AY090567.1 | - | 4 - 5 | 447 | 82 | ||
| BC062378.1 | - | 5 - 6 | 636 | 4978648 | 82 | |
| M84373.1 | - | 5 - 6 | 628 | 82 | ||
| M87039.1 | - | 5 - 6 | 699 | 82 | ||
| M92649.1 | - | 5 - 6 | 635 | 82 | ||
| U43428.1 | - | 5 - 6 | 515 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nos2
Genomic Map