Assay Details
Gene Symbol: Cpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019775.3 | NP_062749.2 | 2 - 3 | 214 | 89 | |
GenBank mRNA | AA048136.1 | - | 2 - 3 | 214 | 89 | |
AB021968.1 | - | 2 - 3 | 162 | 89 | ||
AF164524.1 | - | 2 - 3 | 229 | 89 | ||
AF186188.1 | - | 2 - 3 | 165 | 89 | ||
AK004045.1 | - | 2 - 3 | 187 | 89 | ||
BC089577.1 | - | 2 - 3 | 186 | 6766672 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
