Assay Details
| Gene Symbol: Cpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019775.3 | NP_062749.2 | 3 - 4 | 339 | 73 | |
| GenBank mRNA | AA048136.1 | - | 3 - 4 | 339 | 73 | |
| AB021968.1 | - | 3 - 4 | 287 | 73 | ||
| AF164524.1 | - | 3 - 4 | 354 | 73 | ||
| AF186188.1 | - | 3 - 4 | 290 | 73 | ||
| AK004045.1 | - | 3 - 4 | 312 | 73 | ||
| BC089577.1 | - | 3 - 4 | 311 | 6766672 | 73 | |
More Information
Set Membership: |
|

Cpb2
Genomic Map