Assay Details
Gene Symbol: Cpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019775.3 | NP_062749.2 | 6 - 7 | 652 | 83 | |
GenBank mRNA | AB021968.1 | - | 6 - 7 | 600 | 83 | |
AF164524.1 | - | 6 - 7 | 667 | 83 | ||
AF186188.1 | - | 6 - 7 | 603 | 83 | ||
AK004045.1 | - | 6 - 7 | 625 | 83 | ||
BC089577.1 | - | 6 - 7 | 624 | 6766672 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
