Assay Details
| Gene Symbol: Ankrd13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013806.1 | NP_001013828.1 | 11 - 12 | 1449 | 83 | |
| GenBank mRNA | AK135906.1 | - | 12 - 13 | 1185 | 83 | |
| AK157542.1 | - | 1631 | 83 | |||
| BC065088.1 | - | 10 - 11 | 3643 | 83 | ||
| BC138991.1 | - | 11 - 12 | 1457 | 8862013 | 83 | |
| BC138992.1 | - | 12 - 13 | 1436 | 8862014 | 83 | |
| CF736962.1 | - | 4 - 5 | 445 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd13c
Genomic Map