Assay Details
| Gene Symbol: Ankrd13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013806.1 | NP_001013828.1 | 2 - 3 | 535 | 109 | |
| GenBank mRNA | AK135906.1 | - | 3 - 4 | 271 | 109 | |
| AK157542.1 | - | 763 | 109 | |||
| BC065088.1 | - | 2 - 3 | 548 | 109 | ||
| BC138991.1 | - | 2 - 3 | 543 | 8862013 | 109 | |
| BC138992.1 | - | 3 - 4 | 522 | 8862014 | 109 | |
| BQ562764.1 | - | 2 - 3 | 84 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd13c
Genomic Map