Assay Details
| Gene Symbol: Ankrd13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013806.1 | NP_001013828.1 | 5 - 6 | 772 | 129 | |
| GenBank mRNA | AK135906.1 | - | 6 - 7 | 508 | 129 | |
| BC138991.1 | - | 5 - 6 | 780 | 8862013 | 129 | |
| BC138992.1 | - | 6 - 7 | 759 | 8862014 | 129 | |
| BQ562764.1 | - | 5 - 6 | 321 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd13c
Genomic Map