Assay Details
| Gene Symbol: Ankrd13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013806.1 | NP_001013828.1 | 6 - 7 | 840 | 93 | |
| GenBank mRNA | AK135906.1 | - | 7 - 8 | 576 | 93 | |
| AK157542.1 | - | 1022 | 93 | |||
| BC065088.1 | - | 5 - 6 | 3034 | 93 | ||
| BC138991.1 | - | 6 - 7 | 848 | 8862013 | 93 | |
| BC138992.1 | - | 7 - 8 | 827 | 8862014 | 93 | |
| BQ562764.1 | - | 6 - 7 | 389 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd13c
Genomic Map