Assay Details
| Gene Symbol: Ankrd13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013806.1 | NP_001013828.1 | 7 - 8 | 984 | 162 | |
| GenBank mRNA | AK135906.1 | - | 8 - 9 | 720 | 162 | |
| AK157542.1 | - | 1166 | 162 | |||
| BC065088.1 | - | 6 - 7 | 3178 | 162 | ||
| BC138991.1 | - | 7 - 8 | 992 | 8862013 | 162 | |
| BC138992.1 | - | 8 - 9 | 971 | 8862014 | 162 | |
| BQ562764.1 | - | 7 - 8 | 533 | 162 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd13c
Genomic Map