Assay Details
| Gene Symbol: Ankrd13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013806.1 | NP_001013828.1 | 8 - 9 | 1112 | 112 | |
| GenBank mRNA | AK135906.1 | - | 9 - 10 | 848 | 112 | |
| AK157542.1 | - | 1294 | 112 | |||
| AK190604.1 | - | 1 - 2 | 37 | 112 | ||
| BC065088.1 | - | 7 - 8 | 3306 | 112 | ||
| BC138991.1 | - | 8 - 9 | 1120 | 8862013 | 112 | |
| BC138992.1 | - | 9 - 10 | 1099 | 8862014 | 112 | |
| CF736962.1 | - | 1 - 2 | 108 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd13c
Genomic Map