Assay Details
| Gene Symbol: Gatad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026033.2 | NP_080309.2 | 3 - 4 | 738 | 74 | |
| GenBank mRNA | AB047921.1 | - | 3 - 4 | 719 | 74 | |
| AK012921.1 | - | 3 - 4 | 739 | 74 | ||
| AK078416.1 | - | 3 - 4 | 683 | 74 | ||
| BC019449.1 | - | 3 - 4 | 608 | 4024452 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gatad1
Genomic Map