Assay Details
| Gene Symbol: Arhgap36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081123.1 | NP_001074592.1 | 6 - 7 | 926 | 158 | |
| GenBank mRNA | AB073967.1 | - | 6 - 7 | 793 | 158 | |
| AK048167.1 | - | 6 - 7 | 815 | 158 | ||
| BC090672.1 | - | 6 - 7 | 897 | 30941825 | 158 | |
| BC145645.1 | - | 6 - 7 | 892 | 40130948 | 158 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap36
Genomic Map