Assay Details
| Gene Symbol: Arhgap36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081123.1 | NP_001074592.1 | 9 - 10 | 1403 | 69 | |
| GenBank mRNA | AB073967.1 | - | 9 - 10 | 1270 | 69 | |
| AK048167.1 | - | 9 - 10 | 1292 | 69 | ||
| BC090672.1 | - | 9 - 10 | 1374 | 30941825 | 69 | |
| BC145645.1 | - | 9 - 10 | 1369 | 40130948 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap36
Genomic Map