Assay Details
| Gene Symbol: Wdr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172864.3 | NP_766452.2 | 1 - 2 | 125 | 147 | |
| GenBank mRNA | AK029903.1 | - | 1 - 2 | 126 | 147 | |
| BC158119.1 | - | 1 - 2 | 231 | 9007392 | 148 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC158119.1, NM_172864.3 transcripts.
Set Membership: |
|

Wdr63
Genomic Map