Assay Details
| Gene Symbol: Wdr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172864.3 | NP_766452.2 | 11 - 12 | 1410 | 54 | |
| GenBank mRNA | AK029903.1 | - | 11 - 12 | 1412 | 54 | |
| BC132106.1 | - | 6 - 7 | 848 | 40130383 | 54 | |
| BC132108.1 | - | 6 - 7 | 848 | 40130385 | 54 | |
| BC158119.1 | - | 11 - 12 | 1516 | 9007392 | 54 | |
More Information
Set Membership: |
|

Wdr63
Genomic Map