Assay Details
| Gene Symbol: Wdr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172864.3 | NP_766452.2 | 13 - 14 | 1621 | 137 | |
| GenBank mRNA | AK029903.1 | - | 13 - 14 | 1623 | 137 | |
| BC132106.1 | - | 8 - 9 | 1059 | 40130383 | 137 | |
| BC132108.1 | - | 8 - 9 | 1059 | 40130385 | 137 | |
| BC158119.1 | - | 13 - 14 | 1727 | 9007392 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr63
Genomic Map