Assay Details
| Gene Symbol: Mcee | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028626.1 | NP_082902.1 | 2 - 3 | 418 | 97 | |
| GenBank mRNA | AK003511.1 | - | 2 - 3 | 415 | 97 | |
| BC038157.1 | - | 2 - 3 | 418 | 1479934 | 97 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25176752 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320481 | AK003511.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320481 | BC038157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320481 | NM_028626.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320481 | AK003511.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320481 | BC038157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320481 | NM_028626.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195312 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 169652 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 960896 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748303 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251329 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156538 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 661703 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 33676 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 776145 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832749 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108101 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135506 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112899 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119770 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320481 | AK003511.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320481 | BC038157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320481 | NM_028626.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320481 | AK003511.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320481 | BC038157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320481 | NM_028626.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320481 | AK003511.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320481 | BC038157.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320481 | NM_028626.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320481 | AK003511.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320481 | BC038157.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320481 | NM_028626.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25176767 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119770 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06219095 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01531794 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112899 | - | - |
| Whole Mouse Genome G4122A | A_51_P320481 | AK003511.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320481 | BC038157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320481 | NM_028626.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320481 | AK003511.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320481 | BC038157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320481 | NM_028626.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112899 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119770 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06438447 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06438439 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25176752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25176822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06320113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06635270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06490524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06320128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06438438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176780 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10553915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553914 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553916 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438477_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10553913 | - | - |
| Mouse Genome 430 2.0 Array | 1438477_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1438477_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73724_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73724_PM_at | - | - |
| Mouse Expression Set 430 | 1438477_PM_a_at | - | - |
| Mouse Expression Set 430 | 1438477_a_at | - | - |
| Murine Genome U74v2 Set | 102022_PM_at | - | - |
| Murine Genome U74v2 Set | 102022_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438477_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438477_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438477_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438477_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 715053 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2707494 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2707494 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2707494 | - | - |

Mcee
Genomic Map