Assay Details
| Gene Symbol: Fan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177893.3 | NP_808561.2 | 13 - 14 | 3340 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
DNA repair
nucleotide-excision repair
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
interstrand cross-link repair
nucleic acid phosphodiester bond hydrolysis
nuclease activity
endonuclease activity
exonuclease activity
phosphodiesterase I activity
5'-3' exonuclease activity
hydrolase activity
hydrolase activity, acting on ester bonds
5'-flap endonuclease activity
ubiquitin binding
metal ion binding
flap-structured DNA binding
double-strand break repair via homologous recombination
DNA repair
nucleotide-excision repair
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
interstrand cross-link repair
nucleic acid phosphodiester bond hydrolysis
nuclease activity
endonuclease activity
exonuclease activity
phosphodiesterase I activity
5'-3' exonuclease activity
hydrolase activity
hydrolase activity, acting on ester bonds
5'-flap endonuclease activity
ubiquitin binding
metal ion binding
flap-structured DNA binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2030229 | NM_177893.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2030234 | BC116270.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2030234 | BC116271.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2030234 | AK077918.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2030234 | AK173087.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2030234 | NM_177893.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2030239 | AK173087.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166149 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2030234 | BC116270.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2030234 | BC116271.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2030234 | AK077918.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2030234 | AK173087.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2030234 | NM_177893.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2030239 | AK173087.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2030229 | NM_177893.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166149 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01531726 | - | - |
| Whole Mouse Genome G4122A | A_51_P170772 | AK077918.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06485510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06494326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06485511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06319869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25176498 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10564309 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564312 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564303 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564307 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564311 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564308 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564305 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564304 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564310 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564301 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564302 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10564294 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700029086.mm.1 | NM_177893.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700029086.mm.1 | NM_177893.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700029086.mm.1 | TC0700003412.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4560204 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004560204 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2591619 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2591619 | - | - |
| mouse-6 v1.1 (Illumina) | scl31247.3.4_321-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4560204 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31247.3.4_321-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4560204 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2591619 | - | - |

Fan1
Genomic Map