Assay Details
| Gene Symbol: Fan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177893.3 | NP_808561.2 | 4 - 5 | 1998 | 63 | |
| GenBank mRNA | AK173087.1 | - | 3 - 4 | 1456 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fan1
Genomic Map