Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170591.1 | NP_733479.1 | 1 - 2 | 217 | 75 | |
| GenBank mRNA | AK006062.1 | - | 1 - 2 | 319 | 75 | |
| AK029688.1 | - | 271 | 75 | |||
| AK078279.1 | - | 1 - 2 | 265 | 75 | ||
| AK086412.1 | - | 1 - 2 | 285 | 75 | ||
| AK129134.1 | - | 1 - 2 | 254 | 75 | ||
| AK133721.1 | - | 1 - 2 | 228 | 75 | ||
| AK139895.1 | - | 1 - 2 | 347 | 75 | ||
| AK146477.1 | - | 1 - 2 | 217 | 75 | ||
| AK163792.1 | - | 1 - 2 | 246 | 75 | ||
| BC026743.1 | - | 1 - 2 | 248 | 3964978 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nupl1
Genomic Map