Assay Details
Gene Symbol: Piwil2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021308.1 | NP_067283.1 | 10 - 11 | 1369 | 62 | |
GenBank mRNA | AB032605.1 | - | 10 - 11 | 1369 | 62 | |
AF285586.1 | - | 1 - 2 | 98 | 62 | ||
AK030116.1 | - | 10 - 11 | 1313 | 62 | ||
BC138444.1 | - | 10 - 11 | 1357 | 8861464 | 62 | |
BC145717.1 | - | 10 - 11 | 1357 | 40131020 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF285586.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
