Assay Details
| Gene Symbol: Sdr39u1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082975.1 | NP_001076444.1 | 3 - 4 | 240 | 78 | |
| GenBank mRNA | AK009339.1 | - | 3 - 4 | 220 | 78 | |
| AK049709.1 | - | 3 - 4 | 234 | 78 | ||
| BC087941.1 | - | 3 - 4 | 219 | 30303350 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sdr39u1
Genomic Map