Assay Details
| Gene Symbol: Cmtm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026066.2 | NP_080342.1 | 3 - 4 | 559 | 83 | |
| NM_029390.1 | NP_083666.1 | 2 - 3 | 379 | 83 | ||
| GenBank mRNA | AK005168.1 | - | 2 - 3 | 422 | 83 | |
| AK013666.1 | - | 3 - 4 | 559 | 83 | ||
| AY241867.1 | - | 3 - 4 | 549 | 83 | ||
| BC049665.1 | - | 3 - 4 | 546 | 6533928 | 83 | |
| DQ115983.1 | - | 2 - 3 | 379 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cmtm5
Genomic Map