Assay Details
| Gene Symbol: Zbtb33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020256.2 | NP_064652.2 | 1 - 2 | 288 | 83 | |
| GenBank mRNA | AF097416.1 | - | 1 - 2 | 251 | 83 | |
| AK089423.1 | - | 1 - 2 | 248 | 83 | ||
| BC138647.1 | - | 1 - 2 | 87 | 8861667 | 83 | |
| BC138648.1 | - | 1 - 2 | 87 | 8861668 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF097416.1, AK089423.1, BC138647.1, BC138648.1, NM_020256.2 transcripts.
Set Membership: |
|

Zbtb33
Genomic Map