| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400018823.mm.1 |
NR_045294.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018823.mm.1 |
NR_045294.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018823.mm.1 |
TC1400002871.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400018828.mm.1 |
NR_045294.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018828.mm.1 |
NR_045294.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018828.mm.1 |
TC1400002871.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400018824.mm.1 |
NR_045294.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018824.mm.1 |
NR_045294.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018824.mm.1 |
TC1400002871.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400018822.mm.1 |
NR_045294.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018822.mm.1 |
NR_045294.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018822.mm.1 |
TC1400002871.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400018827.mm.1 |
NR_045294.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018827.mm.1 |
NR_045294.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018827.mm.1 |
TC1400002871.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1456889_at |
- |
- |
| Mouse Expression Set 430 |
1456889_PM_at |
- |
- |
| Murine Genome U74v2 Set |
129300_PM_at |
- |
- |
| Murine Genome U74v2 Set |
129300_at |
- |
- |