Assay Details
| Gene Symbol: Sntn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177624.3 | NP_808292.1 | 1 - 2 | 121 | 93 | |
| GenBank mRNA | AK040281.1 | - | 1 - 2 | 122 | 93 | |
| BC126945.1 | - | 1 - 2 | 122 | 40090204 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sntn
Genomic Map