Assay Details
| Gene Symbol: Whamm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004185.3 | NP_001004185.3 | 4 - 5 | 1105 | 72 | |
| GenBank mRNA | AK140311.1 | - | 4 - 5 | 1115 | 72 | |
| AK162084.1 | - | 6 - 7 | 1110 | 72 | ||
| AK220273.1 | - | 4 - 5 | 1105 | 72 | ||
| BC042749.2 | - | 5 - 6 | 940 | 5322109 | 72 | |
| BC050804.1 | - | 4 - 5 | 568 | 6771347 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Whamm
Genomic Map