Assay Details
| Gene Symbol: Gnrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008145.2 | NP_032171.1 | 3 - 4 | 338 | 62 | |
| GenBank mRNA | AK139480.1 | - | 3 - 4 | 300 | 62 | |
| BC116897.1 | - | 2 - 3 | 310 | 40126216 | 62 | |
| BC116899.1 | - | 2 - 3 | 310 | 40126218 | 62 | |
| CJ059442.1 | - | 3 - 4 | 300 | 62 | ||
| CX763652.1 | - | 3 - 4 | 338 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
regulation of gene expression
male sex determination
negative regulation of immature T cell proliferation
negative regulation of apoptotic process
response to ethanol
response to steroid hormone
regulation of ovarian follicle development
negative regulation of neuron migration
hormone activity
gonadotropin hormone-releasing hormone activity
multicellular organism development
regulation of gene expression
male sex determination
negative regulation of immature T cell proliferation
negative regulation of apoptotic process
response to ethanol
response to steroid hormone
regulation of ovarian follicle development
negative regulation of neuron migration
hormone activity
gonadotropin hormone-releasing hormone activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095716 | BC116897.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095716 | BC116899.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110860 | NM_008145.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110860 | CX763652.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729008 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640441 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177650 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110860 | NM_008145.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110860 | CX763652.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273750 | NM_008145.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273750 | AK139480.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273750 | BC116897.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273750 | BC116899.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273750 | CJ059442.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177650 | - | - |
| Whole Mouse Genome G4122A | A_51_P273750 | NM_008145.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273750 | AK139480.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273750 | BC116897.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273750 | BC116899.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273750 | CJ059442.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133888 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177650 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11359686 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11678220 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11678216 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11359682 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11359688 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11359691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29687026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29686983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11359682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11184789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11310499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11339880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11310505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29687025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29686989 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10416172 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10416173 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10416170 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10416171 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10416174 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10416169 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400007735.mm.1 | NM_008145.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400007735.mm.1 | TC1400000963.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400007736.mm.1 | NM_008145.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400007736.mm.1 | TC1400000963.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 101290_at | - | - |
| Murine Genome U74v2 Set | 101290_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 470940 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160391 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160391 | - | - |
| mouse-6 v1.1 (Illumina) | GI_51093848-S | - | - |
Citations (1)
Altered GABAA receptor-mediated synaptic transmission disrupts the firing of gonadotropin-releasing hormone neurons in male mice under conditions that mimic steroid abuse.
- Authors
- Penatti CA, Davis MC, Porter DM, Henderson LP
- Journal
- J Neurosci - Page No 6497 (Wed May 12 00:00:00 UTC 2010)
- Volume (Year)
- 30 (2010)
- Product Usage
- -

Gnrh1
Genomic Map