Assay Details
| Gene Symbol: Gfra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF002701.1 | - | 2 - 3 | 350 | 70 | |
| AK138512.1 | - | 2 - 3 | 1076 | 70 | ||
| AK149349.1 | - | 2 - 3 | 1080 | 70 | ||
| AK154579.1 | - | 2 - 3 | 1086 | 70 | ||
| BC138561.1 | - | 2 - 3 | 471 | 8861581 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK149349.1 transcripts.
Set Membership: |
|

Gfra2
Genomic Map