Assay Details
| Gene Symbol: Cdk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011074.2 | NP_035204.2 | 1 - 2 | 212 | 68 | |
| GenBank mRNA | AF033655.1 | - | 1 - 2 | 171 | 68 | |
| AK016370.1 | - | 1 - 2 | 154 | 68 | ||
| AK045083.1 | - | 1 - 2 | 213 | 68 | ||
| AK051283.1 | - | 1 - 2 | 186 | 68 | ||
| AK051411.1 | - | 1 - 2 | 155 | 68 | ||
| AK083790.1 | - | 1 - 2 | 155 | 68 | ||
| AK129226.1 | - | 1 - 2 | 39 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk14
Genomic Map