Assay Details
| Gene Symbol: Cdk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011074.2 | NP_035204.2 | 3 - 4 | 494 | 139 | |
| GenBank mRNA | AF033655.1 | - | 3 - 4 | 453 | 139 | |
| AK016370.1 | - | 3 - 4 | 436 | 139 | ||
| AK045083.1 | - | 3 - 4 | 495 | 139 | ||
| AK051283.1 | - | 3 - 4 | 468 | 139 | ||
| AK051411.1 | - | 3 - 4 | 437 | 139 | ||
| AK083269.1 | - | 2 - 3 | 649 | 139 | ||
| AK083790.1 | - | 3 - 4 | 437 | 139 | ||
| AK087398.1 | - | 2 - 3 | 265 | 139 | ||
| AK129226.1 | - | 3 - 4 | 321 | 139 | ||
| AK138437.1 | - | 2 - 3 | 735 | 139 | ||
| AK139386.1 | - | 2 - 3 | 655 | 139 | ||
| AK141535.1 | - | 2 - 3 | 423 | 139 | ||
| AK165149.1 | - | 2 - 3 | 339 | 139 | ||
| BC068134.1 | - | 2 - 3 | 698 | 6834018 | 139 | |
| U62391.1 | - | 2 - 3 | 348 | 139 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk14
Genomic Map