| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500000136.mm.1 |
NM_029141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000136.mm.1 |
NM_029141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000136.mm.1 |
NM_029154.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000136.mm.1 |
NM_029154.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000136.mm.1 |
TC0500000017.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500000137.mm.1 |
NM_029141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000137.mm.1 |
NM_029141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000137.mm.1 |
NM_029154.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000137.mm.1 |
NM_029154.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000137.mm.1 |
TC0500000017.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500000138.mm.1 |
NM_029141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000138.mm.1 |
NM_029141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000138.mm.1 |
NM_029154.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000138.mm.1 |
NM_029154.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000138.mm.1 |
TC0500000017.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500000139.mm.1 |
NM_029141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000139.mm.1 |
NM_029141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000139.mm.1 |
NM_029154.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000139.mm.1 |
NM_029154.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000139.mm.1 |
TC0500000017.mm.1 |
MTA TC |