Assay Details
| Gene Symbol: Tmbim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029141.4 | NP_083417.1 | 4 - 5 | 744 | 63 | |
| NM_029154.1 | NP_083430.1 | 4 - 5 | 552 | 63 | ||
| GenBank mRNA | AK015666.1 | - | 4 - 5 | 744 | 63 | |
| AK015761.1 | - | 4 - 5 | 377 | 63 | ||
| AK019559.1 | - | 4 - 5 | 572 | 63 | ||
| AK161344.1 | - | 2 - 3 | 247 | 63 | ||
| BC115681.2 | - | 4 - 5 | 602 | 40104180 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmbim7
Genomic Map