Assay Details
| Gene Symbol: Npnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029836.2 | NP_001025007.1 | 2 - 3 | 427 | 96 | |
| NM_001287102.1 | NP_001274031.1 | 2 - 3 | 427 | 96 | ||
| NM_001287103.1 | NP_001274032.1 | 2 - 3 | 145 | 96 | ||
| GenBank mRNA | AB059656.1 | - | 2 - 3 | 427 | 96 | |
| AF397007.1 | - | 2 - 3 | 179 | 96 | ||
| AK050484.1 | - | 2 - 3 | 331 | 96 | ||
| AK158764.1 | - | 2 - 3 | 147 | 96 | ||
| AY035898.1 | - | 2 - 3 | 302 | 96 | ||
| BC068308.1 | - | 2 - 3 | 296 | 30472074 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001287103.1 transcripts.
Set Membership: |
|

Npnt
Genomic Map