Assay Details
| Gene Symbol: Npnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029836.2 | NP_001025007.1 | 4 - 5 | 633 | 76 | |
| NM_001287101.1 | NP_001274030.1 | 6 - 7 | 777 | 76 | ||
| NM_001287102.1 | NP_001274031.1 | 5 - 6 | 726 | 76 | ||
| NM_001287103.1 | NP_001274032.1 | 4 - 5 | 351 | 76 | ||
| NM_033525.3 | NP_277060.2 | 5 - 6 | 684 | 76 | ||
| GenBank mRNA | AB059656.1 | - | 4 - 5 | 633 | 76 | |
| AF397007.1 | - | 5 - 6 | 478 | 76 | ||
| AF397008.1 | - | 6 - 7 | 529 | 76 | ||
| AK050484.1 | - | 4 - 5 | 537 | 76 | ||
| AK158764.1 | - | 4 - 5 | 353 | 76 | ||
| AK201444.1 | - | 3 - 4 | 240 | 76 | ||
| AY035898.1 | - | 4 - 5 | 508 | 76 | ||
| AY035899.1 | - | 5 - 6 | 559 | 76 | ||
| BC068308.1 | - | 4 - 5 | 502 | 30472074 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Npnt
Genomic Map