Assay Details
| Gene Symbol: Npnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029836.2 | NP_001025007.1 | 5 - 6 | 748 | 79 | |
| NM_001287101.1 | NP_001274030.1 | 7 - 8 | 892 | 79 | ||
| NM_001287102.1 | NP_001274031.1 | 6 - 7 | 841 | 79 | ||
| NM_001287103.1 | NP_001274032.1 | 5 - 6 | 466 | 79 | ||
| NM_033525.3 | NP_277060.2 | 6 - 7 | 799 | 79 | ||
| GenBank mRNA | AB059656.1 | - | 5 - 6 | 748 | 79 | |
| AF397007.1 | - | 6 - 7 | 593 | 79 | ||
| AF397008.1 | - | 7 - 8 | 644 | 79 | ||
| AK050484.1 | - | 5 - 6 | 652 | 79 | ||
| AK158764.1 | - | 5 - 6 | 468 | 79 | ||
| AY035898.1 | - | 5 - 6 | 623 | 79 | ||
| AY035899.1 | - | 6 - 7 | 674 | 79 | ||
| BC068308.1 | - | 5 - 6 | 617 | 30472074 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Npnt
Genomic Map