Assay Details
| Gene Symbol: Npnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029836.2 | NP_001025007.1 | 7 - 8 | 1010 | 81 | |
| NM_001287101.1 | NP_001274030.1 | 9 - 10 | 1154 | 81 | ||
| NM_001287102.1 | NP_001274031.1 | 8 - 9 | 1103 | 81 | ||
| NM_001287103.1 | NP_001274032.1 | 7 - 8 | 728 | 81 | ||
| NM_033525.3 | NP_277060.2 | 8 - 9 | 1061 | 81 | ||
| GenBank mRNA | AB059656.1 | - | 7 - 8 | 1010 | 81 | |
| AF397007.1 | - | 8 - 9 | 855 | 81 | ||
| AF397008.1 | - | 9 - 10 | 906 | 81 | ||
| AK050484.1 | - | 7 - 8 | 914 | 81 | ||
| AK158764.1 | - | 7 - 8 | 730 | 81 | ||
| AY035898.1 | - | 7 - 8 | 885 | 81 | ||
| AY035899.1 | - | 8 - 9 | 936 | 81 | ||
| BC046642.1 | - | 2 - 3 | 197 | 81 | ||
| BC068308.1 | - | 7 - 8 | 879 | 30472074 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Npnt
Genomic Map