Assay Details
| Gene Symbol: Npnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029836.2 | NP_001025007.1 | 9 - 10 | 1494 | 64 | |
| NM_001287101.1 | NP_001274030.1 | 11 - 12 | 1638 | 64 | ||
| NM_001287102.1 | NP_001274031.1 | 10 - 11 | 1587 | 64 | ||
| NM_001287103.1 | NP_001274032.1 | 9 - 10 | 1212 | 64 | ||
| NM_033525.3 | NP_277060.2 | 10 - 11 | 1545 | 64 | ||
| GenBank mRNA | AB059656.1 | - | 9 - 10 | 1494 | 64 | |
| AF397007.1 | - | 10 - 11 | 1339 | 64 | ||
| AF397008.1 | - | 11 - 12 | 1390 | 64 | ||
| AK050484.1 | - | 9 - 10 | 1398 | 64 | ||
| AK158764.1 | - | 9 - 10 | 1214 | 64 | ||
| AY035898.1 | - | 9 - 10 | 1369 | 64 | ||
| AY035899.1 | - | 10 - 11 | 1420 | 64 | ||
| BC046642.1 | - | 4 - 5 | 681 | 64 | ||
| BC068308.1 | - | 9 - 10 | 1363 | 30472074 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Npnt
Genomic Map