Assay Details
| Gene Symbol: Adh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007409.2 | NP_031435.1 | 2 - 3 | 197 | 72 | |
| GenBank mRNA | AK146094.1 | - | 2 - 3 | 198 | 72 | |
| BC013477.1 | - | 2 - 3 | 197 | 4238555 | 72 | |
| BC054467.1 | - | 2 - 3 | 177 | 6530906 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adh1
Genomic Map