Assay Details
| Gene Symbol: Ngrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031375.4 | NP_113552.3 | 2 - 3 | 313 | 78 | |
| NR_028053.1 | 1 - 2 | 710 | 78 | |||
| GenBank mRNA | AB047544.1 | - | 2 - 3 | 339 | 78 | |
| AK083330.1 | - | 1 - 2 | 706 | 78 | ||
| AK144756.1 | - | 2 - 3 | 309 | 78 | ||
| AK146059.1 | - | 1 - 2 | 688 | 78 | ||
| BC004034.1 | - | 300 | 3495639 | 78 | ||
| CJ154012.1 | - | 2 - 3 | 312 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ngrn
Genomic Map