Assay Details
| Gene Symbol: Negr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039094.3 | NP_001034183.1 | 2 - 3 | 795 | 83 | |
| NM_177274.4 | NP_796248.1 | 2 - 3 | 795 | 83 | ||
| GenBank mRNA | AJ487032.1 | - | 2 - 3 | 415 | 83 | |
| AK030503.1 | - | 2 - 3 | 621 | 83 | ||
| AK035218.1 | - | 2 - 3 | 631 | 83 | ||
| AK122576.2 | - | 1 - 2 | 138 | 83 | ||
| AK142298.1 | - | 2 - 3 | 489 | 83 | ||
| AK147253.1 | - | 2 - 3 | 796 | 83 | ||
| BC146005.1 | - | 2 - 3 | 580 | 40131308 | 83 | |
| BC146007.1 | - | 2 - 3 | 579 | 40131310 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Negr1
Genomic Map