Assay Details
| Gene Symbol: Gjb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271663.1 | NP_001258592.1 | 2 - 3 | 679 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271663.1 transcripts.
Set Membership: |
|

Gjb6
Genomic Map