Assay Details
| Gene Symbol: Gstcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026231.2 | NP_080507.2 | 9 - 10 | 1847 | 102 | |
| GenBank mRNA | AK013967.1 | - | 6 - 7 | 699 | 102 | |
| AK017059.1 | - | 7 - 8 | 1122 | 102 | ||
| AK020021.1 | - | 8 - 9 | 1457 | 102 | ||
| AK029724.1 | - | 9 - 10 | 1848 | 102 | ||
| AK033409.1 | - | 9 - 10 | 1886 | 102 | ||
| BC047136.1 | - | 9 - 10 | 1847 | 6311866 | 102 | |
| BC061240.1 | - | 9 - 10 | 1843 | 6743615 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gstcd
Genomic Map