Assay Details
| Gene Symbol: Gstcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029724.1 | - | 1 - 2 | 158 | 128 | |
| BC061240.1 | - | 1 - 2 | 155 | 6743615 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC061240.1 transcripts.
Set Membership: |
|

Gstcd
Genomic Map