Assay Details
| Gene Symbol: Gstcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026231.2 | NP_080507.2 | 3 - 4 | 1079 | 74 | |
| GenBank mRNA | AK017059.1 | - | 1 - 2 | 354 | 74 | |
| AK020021.1 | - | 2 - 3 | 689 | 74 | ||
| AK029724.1 | - | 3 - 4 | 1078 | 74 | ||
| AK033409.1 | - | 3 - 4 | 1117 | 74 | ||
| AK044441.1 | - | 3 - 4 | 1087 | 74 | ||
| BC047136.1 | - | 3 - 4 | 1079 | 6311866 | 74 | |
| BC061240.1 | - | 3 - 4 | 1075 | 6743615 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017059.1 transcripts.
Set Membership: |
|

Gstcd
Genomic Map